Order Entry
Italy
ContactUsLinkComponent
1-Methyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine 98%
Corrosive
Irritant
1-Methyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine 98%
Catalog # APOSOR16630-250MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
1-Methyl-4-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]piperazine 98%
Catalog # APOSOR16630-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :303.21 g/mol
Melting Pt:125…126 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
 :GHS05
 :GHS07
CAS Number:918524-63-7
Formula:C₁₆H₂₆BN₃O₂
MDL Number:MFCD07437995