tert-Butyl-4-((methylsulphonyl)oxy)piperidine-1-carboxylate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:141699-59-4
:
Formula:C₁₁H₂₁NO₅S
MDL Number:MFCD04116220
:279.3500 g/mol
Storage Temperature:Ambient
Frequently Bought Together