
5-(Morpholin-4-ylmethyl)isoxazole-3-carboxylic acid hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:944450-97-9
:
Formula:C₉H₁₃ClN₂O₄
MDL Number:MFCD08277061
:248.67 g/mol
Melting Pt:154…160 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together