
(5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:112741-49-8
:
Formula:C₂₁H₂₃NO₄
MDL Number:MFCD00074955
:353.418 g/mol
Melting Pt:206 °C (dec.)
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together