
5-Methoxynicotinic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:20826-03-3
:
Formula:C7H7NO3
MDL Number:MFCD01603648
:153.1400 g/mol
Melting Pt:127…131 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together