
3,4-Dibromothiophene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3141-26-2
:
Formula:C₄H₂Br₂S
MDL Number:MFCD00005465
:241.93 g/mol
Boiling Pt:221…222 °C
Melting Pt:44320 °C
Density:2.188 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:221-546-8
:GHS07
Storage Temperature:Ambient
Frequently Bought Together