
4-tert-Butyl-1-(bromomethyl)benzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18880-00-7
:
Formula:C₁₁H₁₅Br
MDL Number:MFCD00000180
:227.14 g/mol
Boiling Pt:93…94 °C/1.5mmHg
Melting Pt:44538 °C
Density:1.236 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together