Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-1-Boc-2-(aminomethyl)pyrrolidine
(S)-1-Boc-2-(aminomethyl)pyrrolidine
(S)-1-Boc-2-(aminomethyl)pyrrolidine
(S)-1-Boc-2-(aminomethyl)pyrrolidine
APOSOR15683-5G
:
Apollo Scientific
:
(S)-1-Boc-2-(aminomethyl)pyrrolidine
APOSOR15683-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
112 °C/ 0.75mmHg
:
Refrigerator
:
2735
:
8,III
:
200.28 g/mol
:
GHS05
:
GHS07
:
119020-01-8
:
C₁₀H₂₀N₂O₂
:
MFCD03419257