Order Entry
Italy
ContactUsLinkComponent
 
Irritant
5-Methylimidazo[1,2-a]pyridine
Catalog # APOSOR15456-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Methylimidazo[1,2-a]pyridine
Catalog # APOSOR15456-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :132.1600 g/mol
Boiling Pt:96 °C/0.7mmHg
 :GHS07
Storage Temperature:Ambient
CAS Number:933-69-7
Formula:C₈H₈N₂
MDL Number:MFCD06200991