Order Entry
Italy
ContactUsLinkComponent
tert-Butyl-2-amino-3-cyano-4,7-dihydrothieno[2,3-c]pyridine-6(5H)-carboxylate
Irritant
tert-Butyl-2-amino-3-cyano-4,7-dihydrothieno[2,3-c]pyridine-6(5H)-carboxylate
Catalog # APOSOR15388-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl-2-amino-3-cyano-4,7-dihydrothieno[2,3-c]pyridine-6(5H)-carboxylate
Catalog # APOSOR15388-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :279.36 g/mol
Melting Pt:193…195 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:150986-83-7
Formula:C₁₃H₁₇N₃O₂S
MDL Number:MFCD05664037