3'-Nitro[1,1'-biphenyl]-2-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:209863-08-1
:
Formula:C₁₃H₉NO₃
MDL Number:MFCD06803021
:227.22 g/mol
Melting Pt:100…102 °C
Storage Temperature:Ambient
Frequently Bought Together