Order Entry
Italy
ContactUsLinkComponent
Ethyl-2-(4-chlorophenyl)-4-hydroxy-1,3-thiazole-5-carboxylate
Ethyl-2-(4-chlorophenyl)-4-hydroxy-1,3-thiazole-5-carboxylate
Catalog # APOSOR1528-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2-(4-chlorophenyl)-4-hydroxy-1,3-thiazole-5-carboxylate
Catalog # APOSOR1528-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:137…139 °C
 :283.74 g/mol
Storage Temperature:Ambient
CAS Number:263016-08-6
Formula:C₁₂H₁₀ClNO₃S
MDL Number:MFCD00816787