Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde 98%
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde 98%
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde 98%
APOSOR15246-1G
:
Apollo Scientific
:
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde 98%
APOSOR15246-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
192.26 g/mol
:
GHS07
:
Ambient
:
85943-26-6
:
C₁₂H₁₆O₂
:
MFCD06246086