Order Entry
Italy
ContactUsLinkComponent
Pyrrolo[1,2-c]pyrimidine-3-carboxylic acid hydrochloride hydrate
Pyrrolo[1,2-c]pyrimidine-3-carboxylic acid hydrochloride hydrate
Catalog # APOSOR15244-1G
Supplier:  Apollo Scientific
CAS Number:  
Pyrrolo[1,2-c]pyrimidine-3-carboxylic acid hydrochloride hydrate
Catalog # APOSOR15244-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :216.63 g/mol
Melting Pt:243…245 °C
Storage Temperature:Ambient
Formula:C₈H₉ClN₂O₃
MDL Number:MFCD08443998