Order Entry
Italy
ContactUsLinkComponent
Methyl-3-hydroxy-4-methoxybenzoate
Irritant
Methyl-3-hydroxy-4-methoxybenzoate
Catalog # APOSOR15107-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-3-hydroxy-4-methoxybenzoate
Catalog # APOSOR15107-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.1733 g/mol
Melting Pt:64…67 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:6702-50-7
Formula:C₉H₁₀O₄
MDL Number:MFCD01321262