
tert-Butyl-2,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:924869-27-2
:
Formula:C₁₁H₁₇N₃O₂
MDL Number:MFCD08689720
:223.28 g/mol
Melting Pt:127…128 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together