
Methyl-1,3-benzodioxole-5-carboxylate
Catalog # APOSOR14837-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:180.16 g/mol
Melting Pt:50…52 °C
:GHS07
Storage Temperature:Ambient
CAS Number:326-56-7
Formula:C₉H₈O₄
MDL Number:MFCD00030273