Order Entry
Italy
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine 98%
Irritant
N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine 98%
Catalog # APOSOR14557-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-(tert-Butoxycarbonyl)-4-bromo-D-phenylalanine 98%
Catalog # APOSOR14557-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :344.21 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:79561-82-3
Formula:C₁₄H₁₈BrNO₄
MDL Number:MFCD00270359