Order Entry
Italy
ContactUsLinkComponent
2-(5-Formyl-2-methoxyphenoxy)-2-phenylacetic acid
2-(5-Formyl-2-methoxyphenoxy)-2-phenylacetic acid
Catalog # APOSOR14275-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(5-Formyl-2-methoxyphenoxy)-2-phenylacetic acid
Catalog # APOSOR14275-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:159…161 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:885949-43-9
Formula:C₁₆H₁₄O₅