Order Entry
Italy
ContactUsLinkComponent
4-Acetyl-8-benzyl-1-thia-4,8-diazaspiro[4.5]decane-3-carboxylic acid hydrochloride
Irritant
4-Acetyl-8-benzyl-1-thia-4,8-diazaspiro[4.5]decane-3-carboxylic acid hydrochloride
Catalog # APOSOR14236-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Acetyl-8-benzyl-1-thia-4,8-diazaspiro[4.5]decane-3-carboxylic acid hydrochloride
Catalog # APOSOR14236-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :370.9 g/mol
Melting Pt:251…253 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1030253-99-6
Formula:C₁₇H₂₃ClN₂O₃S
MDL Number:MFCD06795478