
2,2-Dimethoxy-1-piperidino-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16695-59-3
:
Formula:C₉H₁₇NO₃
MDL Number:MFCD08689667
:187.24 g/mol
Boiling Pt:72 °C/0.01mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together