Order Entry
Italy
ContactUsLinkComponent
Pyrazolo[1,5-a]pyridine-2-carboxylic acid 97+%
Irritant
Pyrazolo[1,5-a]pyridine-2-carboxylic acid 97+%
Catalog # APOSOR1408-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Pyrazolo[1,5-a]pyridine-2-carboxylic acid 97+%
Catalog # APOSOR1408-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :162.1500 g/mol
Melting Pt:191…193 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:63237-88-7
Formula:C₈H₆N₂O₂
MDL Number:MFCD06739057