
8-Amino-4H-1,4-benzoxazin-3-one
Catalog # APOSOR13819-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:164.17 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:321126-82-3
Formula:C₈H₈N₂O₂
MDL Number:MFCD09998373