
2-Morpholin-4-yl-2-pyridin-3-ylethylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:410544-52-4
:
Formula:C₁₁H₁₇N₃O
MDL Number:MFCD01631945
:207.28 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
:GHS05
Frequently Bought Together