Order Entry
Italy
ContactUsLinkComponent
2-Methyl-6-oxo-1,6-dihydro-3,4'-bipyridine-5-carboxamide 98+%
2-Methyl-6-oxo-1,6-dihydro-3,4'-bipyridine-5-carboxamide 98+%
Catalog # APOSOR12587-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Methyl-6-oxo-1,6-dihydro-3,4'-bipyridine-5-carboxamide 98+%
Catalog # APOSOR12587-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :229.24 g/mol
Storage Temperature:Ambient
CAS Number:80047-24-1
Formula:C₁₂H₁₁N₃O₂
MDL Number:MFCD09800601