
2-Morpholin-4-yl-1,3-thiazole-4-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:906353-04-6
:
Formula:C₈H₁₀N₂O₃S
MDL Number:MFCD09702410
:214.24 g/mol
Melting Pt:175…177 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together