Order Entry
Italy
ContactUsLinkComponent
4-(6-Chloropyrazin-2-yl)morpholine 98%
Irritant
4-(6-Chloropyrazin-2-yl)morpholine 98%
Catalog # APOSOR11963-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-(6-Chloropyrazin-2-yl)morpholine 98%
Catalog # APOSOR11963-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:720693-19-6
Formula:C₈H₁₀ClN₃O