Order Entry
Italy
ContactUsLinkComponent
 
Ethyl-7-methylimidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSOR11957-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-7-methylimidazo[1,2-a]pyridine-2-carboxylate
Catalog # APOSOR11957-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :204.2300 g/mol
Storage Temperature:Ambient
CAS Number:70705-33-8
Formula:C₁₁H₁₂N₂O₂
MDL Number:MFCD02187526