Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 8-bromo-4-hydroxyquinoline-3-carboxylate
Ethyl-8-bromo-4-hydroxyquinoline-3-carboxylate
Ethyl-8-bromo-4-hydroxyquinoline-3-carboxylate
APOSOR11682-1G
:
Apollo Scientific
:
Ethyl-8-bromo-4-hydroxyquinoline-3-carboxylate
APOSOR11682-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
296.1167 g/mol
:
256…257 °C
:
GHS07
:
Ambient
:
35975-57-6
:
C₁₂H₁₀BrNO₃
:
MFCD00173380