
2',5'-Dimethoxyacetophenone 99+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1201-38-3
:
Formula:C₁₀H₁₂O₃
MDL Number:MFCD00008728
:180.2 g/mol
Boiling Pt:154…156 °C/10mmHg
Melting Pt:18…20 °C
Density:1.139 g/cm³ (25 °C)
Flash Pt:>113 °C
EINECS:214-858-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together