
6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:33050-32-7
:
Formula:C₅H₃ClN₄O
MDL Number:MFCD01219639
:170.56 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together