
4-Chloro-6-morpholin-4-ylpyrimidin-2-ylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:339016-18-1
:
Formula:C₈H₁₁ClN₄O
MDL Number:MFCD00664697
:214.66 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together