Ethyl-1-pyrimidin-2-ylpiperidine-4-carboxylate 95+%
Catalog # APOSOR111002-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:235.2800 g/mol
Storage Temperature:Ambient
CAS Number:111247-60-0
Formula:C₁₂H₁₇N₃O₂