Order Entry
Italy
ContactUsLinkComponent
6,7-Dihydro-4H-pyrano[4,3-d]thiazol-2-amine
6,7-Dihydro-4H-pyrano[4,3-d]thiazol-2-amine
Catalog # APOSOR110862-1G
Supplier:  Apollo Scientific
CAS Number:  
6,7-Dihydro-4H-pyrano[4,3-d]thiazol-2-amine
Catalog # APOSOR110862-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :156.205 g/mol
Storage Temperature:Refrigerator
CAS Number:259810-12-3
Formula:C₆H₈N₂OS
MDL Number:MFCD08690160