
4-(8-methylimidazo[1,2-a]pyridin-2-yl)aniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:365565-88-4
:
Formula:C₁₄H₁₃N₃
MDL Number:MFCD04971851
:223.279 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together