6-(tert-Butoxycarbonyl)-2-(1H-pyrrol-1-yl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1215764-56-9
:
Formula:C₁₇H₂₀N₂O₄S
MDL Number:MFCD11100094
:348.42 g/mol
Storage Temperature:Ambient
Frequently Bought Together