Order Entry
Italy
ContactUsLinkComponent
4-Bromo-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole
4-Bromo-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole
Catalog # APOSOR110575-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-1-tetrahydro-2H-pyran-4-yl-1H-pyrazole
Catalog # APOSOR110575-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 4-Bromo-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazole
 :
 :231.1 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1040377-02-3
Formula:C₈H₁₁BrN₂O
MDL Number:MFCD11505042