Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid
APOSOR110102-1G
:
Apollo Scientific
:
7-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid
APOSOR110102-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
241.05 g/mol
:
234…236 °C
:
GHS07
:
Ambient
:
1019018-46-2
:
C₈H₅BrN₂O₂
:
MFCD22989340