Order Entry
Italy
ContactUsLinkComponent
3-(1H-Pyrazol-1-ylmethyl)benzaldehyde
3-(1H-Pyrazol-1-ylmethyl)benzaldehyde
Catalog # APOSOR110014-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(1H-Pyrazol-1-ylmethyl)benzaldehyde
Catalog # APOSOR110014-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:78425-11-3
Formula:C₁₁H₁₀N₂O
MDL Number:MFCD08059836