
(Methoxymethyl)triphenylphosphonium bromide 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:33670-32-5
:
Formula:C₂₀H₂₀BrOP
MDL Number:MFCD00075442
:387.2500 g/mol
Melting Pt:230 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together