Order Entry
Italy
ContactUsLinkComponent
2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1H-isoindole-1,3(2H)-dione 98%
Irritant
2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1H-isoindole-1,3(2H)-dione 98%
Catalog # APOSOR10581-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1H-isoindole-1,3(2H)-dione 98%
Catalog # APOSOR10581-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :363.22 g/mol
Melting Pt:166…171 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:138500-87-5
Formula:C₂₁H₂₂BNO₄
MDL Number:MFCD02179489