Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Boc(2-aminomethyl)morpholine
4-Boc(2-aminomethyl)morpholine
4-Boc(2-aminomethyl)morpholine
4-Boc(2-aminomethyl)morpholine
APOSOR103659-1G
:
Apollo Scientific
:
4-Boc(2-aminomethyl)morpholine
APOSOR103659-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
216.2774 g/mol
:
GHS07
:
Refrigerator
:
140645-53-0
:
C₁₀H₂₀N₂O₃
:
MFCD06410633