Order Entry
Italy
ContactUsLinkComponent
3-Bromo-2-thiophenesulphonyl chloride 90%
3-Bromo-2-thiophenesulphonyl chloride 90%
Catalog # APOSOR103634-50MG
Supplier:  Apollo Scientific
CAS Number:  
3-Bromo-2-thiophenesulphonyl chloride 90%
Catalog # APOSOR103634-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
Melting Pt:39…41.5 °C
Storage Temperature:Ambient
UN:2923
ADR:8+6.1,II
 :GHS05
 :GHS06
CAS Number:170727-02-3
Formula:C₄H₂BrClO₂S₂
MDL Number:MFCD08059484