

2,4-Dimethoxyphenyl isothiocyanate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:33904-03-9
:
Formula:C₉H₉NO₂S
MDL Number:MFCD00041058
:195.24 g/mol
Boiling Pt:166…167 °C/6mmHg
Melting Pt:51…53 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2923
ADR:8+6.1,III
:GHS05
:GHS07
Frequently Bought Together