
2,2-Dichlorocyclopropanecarboxamide
Catalog # APOSOR0348-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:154 g/mol
Melting Pt:148…149 °C
:GHS07
Storage Temperature:Ambient
CAS Number:75885-60-8
Formula:C₄H₅Cl₂NO
MDL Number:MFCD00218854