Order Entry
Italy
ContactUsLinkComponent
tert-Butyl (piperazin-1-yl)acetate dihydrochloride 97%
Irritant
tert-Butyl (piperazin-1-yl)acetate dihydrochloride 97%
Catalog # APOSOR0238-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
tert-Butyl (piperazin-1-yl)acetate dihydrochloride 97%
Catalog # APOSOR0238-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :273.20 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:827614-56-2
Formula:C₁₀H₂₀N₂O₂.2HCl
MDL Number:MFCD04972649