Ethyl-3-({[(2-chloroethyl)amino]carbonyl}amino)benzoate
Catalog # APOSOR01974-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:270.72 g/mol
Storage Temperature:Ambient
CAS Number:803729-93-3
Formula:C₁₂H₁₅ClN₂O₃
MDL Number:MFCD09836176