
4-(3-Chlorophenyl)-1-butene
Catalog # APOSOR01885-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:166.65 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:91426-46-9
Formula:C₁₀H₁₁Cl
MDL Number:MFCD01319546