Order Entry
Italy
ContactUsLinkComponent
5-Pyridin-2-yl-1H-pyrazole-3-carboxylic acid 97%
Irritant
5-Pyridin-2-yl-1H-pyrazole-3-carboxylic acid 97%
Catalog # APOSOR01503-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Pyridin-2-yl-1H-pyrazole-3-carboxylic acid 97%
Catalog # APOSOR01503-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :189.1708 g/mol
Melting Pt:300 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:374064-02-5
Formula:C9H7N3O2
MDL Number:MFCD05678901