Order Entry
Italy
ContactUsLinkComponent
1,5-Diphenyl-1H-pyrazole-3-carboxylic acid
Irritant
1,5-Diphenyl-1H-pyrazole-3-carboxylic acid
Catalog # APOSOR01492-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,5-Diphenyl-1H-pyrazole-3-carboxylic acid
Catalog # APOSOR01492-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :264.29 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:13599-22-9
Formula:C₁₆H₁₂N₂O₂
MDL Number:MFCD01427273